siRNA Details
| Gene Symbol: LOC100128554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130748.1 | 7 | 1156 | |||
| GenBank mRNA | BC035235.1 | - | 8 | 1332 | ||
| BC039370.1 | - | 7 | 1156 | |||
| BC118583.1 | - | 7 | 700 | 40085030 | ||
| BC121801.1 | - | 8 | 939 | |||

LOC100128554
Genomic Map