siRNA Details
Gene Symbol: LINC00887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054628.1 | - | 6 | 1421 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000429578.5 | - | 6 | 1417 | Chr. 3 - 194299635 - 194312803 [-] on Build GRCh38 |
