siRNA Details
Gene Symbol: LOC152225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026934.1 | 1 | 76 | |||
GenBank mRNA | AK090588.1 | - | 1 | 76 | ||
BG249468.1 | - | 1 | 77 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000465215.1 | - | 1 | 76 | Chr. 3 - 101940859 - 101997926 [+] on Build GRCh38 |
