siRNA Details
| Gene Symbol: LOC101926911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110104.1 | 3 | 1084 | |||
| GenBank mRNA | AK055351.1 | - | 3 | 1084 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926911| Gene Symbol: LOC101926911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110104.1 | 3 | 1084 | |||
| GenBank mRNA | AK055351.1 | - | 3 | 1084 | ||