siRNA Details
| Gene Symbol: LOC101926911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110104.1 | 3 | 915 | |||
| GenBank mRNA | AK055351.1 | - | 3 | 915 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501381.3 | - | 3 | 915 | Chr. 15 - 91022622 - 91031140 [+] on Build GRCh38 | |

LOC101926911
Genomic Map