siRNA Details
| Gene Symbol: LOC105371730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136413.1 | 2 | 821 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000398832.2 | - | 2 | 821 | Chr. 17 - 32141226 - 32143135 [-] on Build GRCh38 | |

LOC105371730
Genomic Map