siRNA Details
Gene Symbol: LOC105371730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136413.1 | 2 | 1272 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000398832.2 | - | 2 | 1272 | Chr. 17 - 32141226 - 32143135 [-] on Build GRCh38 |
