siRNA Details
| Gene Symbol: ESR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX247957.1 | - | 3 | 1259 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000359491.1 | - | 3 | 1259 | Chr. 14 - 64335835 - 64338599 [-] on Build GRCh38 | |

ESR2
Genomic Map