siRNA Details
| Gene Symbol: LOC101927588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125420.1 | 3 | 768 | |||
| GenBank mRNA | BC015129.1 | - | 3 | 772 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000523703.1 | - | 3 | 768 | Chr. 8 - 124192671 - 124247398 [-] on Build GRCh38 | |

LOC101927588
Genomic Map