siRNA Details
| Gene Symbol: LINC01588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126496.1 | 1 | 19 | |||
| NR_126497.1 | 1 | 163 | ||||
| NR_126498.1 | 1 | 1020 | ||||
| NR_126500.1 | 1 | 163 | ||||
| GenBank mRNA | AK090420.1 | - | 1 | 19 | ||
| AK091232.1 | - | 1 | 1019 | |||
| BC128083.1 | - | 1 | 742 | 40075904 | ||
| BC144351.1 | - | 1 | 162 | 9052871 | ||

LINC01588
Genomic Map