siRNA Details
| Gene Symbol: LOC153684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015447.1 | 2 | 1716 | |||
| GenBank mRNA | AK002146.1 | - | 3 | 678 | ||
| AK123995.1 | - | 2 | 1758 | |||
| BC101382.1 | - | 2 | 1768 | |||
| BC101383.2 | - | 2 | 1460 | |||
| BC101384.1 | - | 2 | 1717 | |||
| BC101385.2 | - | 2 | 1716 | |||

LOC153684
Genomic Map