siRNA Details
Gene Symbol: LOC153684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015447.1 | 2 | 1978 | |||
GenBank mRNA | AK002146.1 | - | 3 | 940 | ||
AK123995.1 | - | 2 | 2020 | |||
BC101382.1 | - | 2 | 2030 | |||
BC101383.2 | - | 2 | 1722 | |||
BC101384.1 | - | 2 | 1979 | |||
BC101385.2 | - | 2 | 1978 |
