siRNA Details
Gene Symbol: LOC153684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015447.1 | 2 | 2002 | |||
GenBank mRNA | AK002146.1 | - | 3 | 964 | ||
AK123995.1 | - | 2 | 2044 | |||
BC101382.1 | - | 2 | 2054 | |||
BC101383.2 | - | 2 | 1746 | |||
BC101384.1 | - | 2 | 2003 | |||
BC101385.2 | - | 2 | 2002 |
