siRNA Details
Gene Symbol: LINC00269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103715.1 | 5 | 1285 | |||
GenBank mRNA | AK090929.1 | - | 5 | 1285 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000399711.1 | - | 5 | 1285 | Chr. X - 69179557 - 69209924 [+] on Build GRCh38 |
