siRNA Details
Gene Symbol: LOC100506688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104614.1 | 2 | 2299 | |||
NR_104615.1 | 2 | 2233 | ||||
GenBank mRNA | AK126199.1 | - | 2 | 2267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000399869.1 | - | 2 | 2267 | Chr. 5 - 987180 - 997308 [-] on Build GRCh38 |
