siRNA Details
Gene Symbol: AATBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093410.1 | - | 2, 3 | 789 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448247.5 | - | 2, 3 | 789 | Chr. 21 - 43805758 - 43810361 [-] on Build GRCh38 |
