siRNA Details
| Gene Symbol: LINC00189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027072.2 | 2 | 928 | |||
| GenBank mRNA | AF490769.1 | - | 4 | 603 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420364.1 | - | 2 | 928 | Chr. 21 - 29193494 - 29288205 [+] on Build GRCh38 | |

LINC00189
Genomic Map