siRNA Details
Gene Symbol: LINC01356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103746.1 | 3, 4 | 1160 | |||
GenBank mRNA | AK093435.1 | - | 3, 4 | 1160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000401018.5 | - | 3, 4 | 1160 | Chr. 1 - 112820174 - 112850643 [-] on Build GRCh38 |
