siRNA Details
| Gene Symbol: LINC01356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103746.1 | 1 | 53 | |||
| GenBank mRNA | AK093435.1 | - | 1 | 53 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000401018.5 | - | 1 | 53 | Chr. 1 - 112820174 - 112850643 [-] on Build GRCh38 | |

LINC01356
Genomic Map