siRNA Details
| Gene Symbol: LINC00955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC131584.1 | - | 2, 3 | 327 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000502775.1 | - | 2, 3 | 327 | Chr. 4 - 3576934 - 3590711 [+] on Build GRCh38 | |

LINC00955
Genomic Map