siRNA Details
| Gene Symbol: LINC00955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040045.1 | 3 | 1869 | |||
| GenBank mRNA | AK092743.1 | - | 3 | 1867 | ||
| AX747702.1 | - | 3 | 1867 | |||
| BC131584.1 | - | 4 | 516 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000502775.1 | - | 4 | 516 | Chr. 4 - 3576934 - 3590711 [+] on Build GRCh38 | |

LINC00955
Genomic Map