siRNA Details
Gene Symbol: LOC100506302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126344.1 | 3 | 388 | |||
GenBank mRNA | AK091942.1 | - | 1 | 1507 | ||
BQ055858.1 | - | 3 | 359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000460022.6 | - | 3 | 359 | Chr. 7 - 155642264 - 155644377 [-] on Build GRCh38 |
