siRNA Details
| Gene Symbol: LOC100506302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126344.1 | 1, 2 | 197 | |||
| GenBank mRNA | BQ055858.1 | - | 1, 2 | 168 | ||
| DB170406.1 | - | 1, 2 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506302| Gene Symbol: LOC100506302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126344.1 | 1, 2 | 197 | |||
| GenBank mRNA | BQ055858.1 | - | 1, 2 | 168 | ||
| DB170406.1 | - | 1, 2 | 197 | |||