siRNA Details
| Gene Symbol: LOC100506302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126344.1 | 3 | 498 | |||
| GenBank mRNA | AK091942.1 | - | 1 | 1617 | ||
| BQ055858.1 | - | 3 | 469 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000460022.6 | - | 3 | 469 | Chr. 7 - 155642264 - 155644377 [-] on Build GRCh38 | |

LOC100506302
Genomic Map