siRNA Details
| Gene Symbol: LOC285095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135766.1 | 2, 3 | 269 | |||
| NR_135767.1 | 2, 3 | 229 | ||||
| NR_135768.1 | 2, 3 | 168 | ||||
| GenBank mRNA | AA250751.1 | - | 2, 3 | 195 | ||
| BX108328.1 | - | 2, 3 | 235 | |||
| HY129633.1 | - | 2, 3 | 270 | |||

LOC285095
Genomic Map