siRNA Details
Gene Symbol: MIF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038911.1 | 3 | 1405 | |||
GenBank mRNA | BC036909.1 | - | 3 | 1405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000406213.1 | - | 3 | 1405 | Chr. 22 - 23894426 - 23898930 [-] on Build GRCh38 |
