siRNA Details
| Gene Symbol: LOC100289094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC025775.1 | - | 1 | 16 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000407635.2 | - | 3 | 2464 | Chr. 2 - 240686334 - 240690414 [+] on Build GRCh38 | |

LOC100289094
Genomic Map