siRNA Details
| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CA442809.1 | - | 1 | 328 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000396799.3 | - | 1 | 395 | Chr. 12 - 6667234 - 6689510 [-] on Build GRCh38 | |

ZNF384
Genomic Map