siRNA Details
| Gene Symbol: LOC101928254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110182.1 | 5 | 768 | |||
| NR_110183.1 | 4 | 581 | ||||
| GenBank mRNA | CV808846.1 | - | 4 | 564 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420224.1 | - | 5 | 768 | Chr. 7 - 125229579 - 125264291 [-] on Build GRCh38 | |

LOC101928254
Genomic Map