siRNA Details
| Gene Symbol: FLJ35934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104343.1 | 1 | 1308 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000577684.1 | - | 1 | 1308 | Chr. 17 - 18411159 - 18414380 [+] on Build GRCh38 | |

FLJ35934
Genomic Map