siRNA Details
Gene Symbol: FLJ35934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104343.1 | 1 | 1336 | |||
GenBank mRNA | AK093253.1 | - | 1 | 1334 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000577684.1 | - | 1 | 1336 | Chr. 17 - 18411159 - 18414380 [+] on Build GRCh38 |
