siRNA Details
Gene Symbol: PPP3CB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132103.1 | 2 | 367 | |||
NR_132104.1 | 2 | 367 | ||||
GenBank mRNA | BU150385.1 | - | 1 | 161 | ||
DA883449.1 | - | 1 | 315 | |||
DB134060.1 | - | 2 | 368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000422977.1 | - | 2 | 594 | Chr. 10 - 73495525 - 73496778 [+] on Build GRCh38 |
