siRNA Details
Gene Symbol: LOC101928111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135575.1 | 1, 2 | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000409526.1 | - | 1, 2 | 106 | Chr. 2 - 239401436 - 239402357 [+] on Build GRCh38 |
