siRNA Details
| Gene Symbol: LOC101928111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135575.1 | 2 | 361 | |||
| NR_135576.1 | 2 | 510 | ||||
| NR_135577.1 | 2 | 393 | ||||
| GenBank mRNA | BM704421.1 | - | 2 | 393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928111| Gene Symbol: LOC101928111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135575.1 | 2 | 361 | |||
| NR_135576.1 | 2 | 510 | ||||
| NR_135577.1 | 2 | 393 | ||||
| GenBank mRNA | BM704421.1 | - | 2 | 393 | ||