siRNA Details
| Gene Symbol: LOC101927604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110681.1 | 3 | 714 | |||
| GenBank mRNA | BC032911.1 | - | 3 | 719 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439341.1 | - | 3 | 714 | Chr. 1 - 230874846 - 230879015 [+] on Build GRCh38 | |

LOC101927604
Genomic Map