siRNA Details
| Gene Symbol: LINC00473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026860.1 | 2 | 1708 | |||
| GenBank mRNA | BC008632.1 | - | 2 | 1708 | ||
| BC039614.1 | - | 2 | 1708 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455853.1 | - | 2 | 1708 | Chr. 6 - 165939456 - 165988039 [-] on Build GRCh38 | |

LINC00473
Genomic Map