siRNA Details
| Gene Symbol: LINC00473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX647106.1 | - | 2 | 1237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000584911.1 | - | 2 | 1237 | Chr. 6 - 165948223 - 165988048 [-] on Build GRCh38 | |

LINC00473
Genomic Map