siRNA Details
| Gene Symbol: TH2LCRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132125.1 | 3 | 294 | |||
| NR_132126.1 | 2 | 279 | ||||
| GenBank mRNA | AA470013.1 | - | 2 | 279 | ||
| BC042122.1 | - | 3 | 294 | |||
| BX095327.1 | - | 2 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TH2LCRR| Gene Symbol: TH2LCRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132125.1 | 3 | 294 | |||
| NR_132126.1 | 2 | 279 | ||||
| GenBank mRNA | AA470013.1 | - | 2 | 279 | ||
| BC042122.1 | - | 3 | 294 | |||
| BX095327.1 | - | 2 | 278 | |||