siRNA Details
| Gene Symbol: LOC100506100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046240.1 | 1, 2 | 169 | |||
| GenBank mRNA | BX090437.1 | - | 1, 2 | 163 | ||
| EB386762.1 | - | 1, 2 | 169 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443631.1 | - | 1, 2 | 186 | Chr. 9 - 128724445 - 128733194 [+] on Build GRCh38 | |

LOC100506100
Genomic Map