siRNA Details
| Gene Symbol: LINC00630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038988.1 | 8 | 996 | |||
| GenBank mRNA | BC040959.1 | - | 8 | 984 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000440496.5 | - | 8 | 984 | Chr. X - 102769179 - 102885406 [+] on Build GRCh38 | |

LINC00630
Genomic Map