siRNA Details
| Gene Symbol: LOC101928012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110158.1 | 8 | 1024 | |||
| GenBank mRNA | BC041883.1 | - | 8 | 1027 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453459.1 | - | 8 | 1027 | Chr. 7 - 112622378 - 112708080 [+] on Build GRCh38 | |

LOC101928012
Genomic Map