siRNA Details
Gene Symbol: LOC646626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125723.1 | - | 4 | 363 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000427819.5 | - | 4 | 363 | Chr. 1 - 85276715 - 85399963 [+] on Build GRCh38 |
