siRNA Details
| Gene Symbol: LOC105373185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135616.1 | 2 | 202 | |||
| GenBank mRNA | BE613553.1 | - | 2 | 202 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000452501.1 | - | 2 | 202 | Chr. X - 41275739 - 41276778 [+] on Build GRCh38 | |

LOC105373185
Genomic Map