siRNA Details
Gene Symbol: LINC01208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109968.1 | 3 | 500 | |||
GenBank mRNA | BX094192.1 | - | 3 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434969.1 | - | 3 | 500 | Chr. 3 - 176604143 - 176635532 [-] on Build GRCh38 |
