siRNA Details
| Gene Symbol: LOC100130417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026874.2 | 4 | 853 | |||
| NR_122045.1 | 4 | 793 | ||||
| GenBank mRNA | BC025320.1 | - | 1 | 1200 | ||
| BX108591.1 | - | 1 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130417| Gene Symbol: LOC100130417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026874.2 | 4 | 853 | |||
| NR_122045.1 | 4 | 793 | ||||
| GenBank mRNA | BC025320.1 | - | 1 | 1200 | ||
| BX108591.1 | - | 1 | 133 | |||