siRNA Details
| Gene Symbol: LOC101927356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110076.1 | 5 | 1606 | |||
| NR_110077.1 | 4 | 1522 | ||||
| GenBank mRNA | AK055932.1 | - | 5 | 1606 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439234.5 | - | 5 | 1606 | Chr. 7 - 82009177 - 82029955 [+] on Build GRCh38 | |

LOC101927356
Genomic Map