siRNA Details
| Gene Symbol: ENTPD3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040100.1 | 1, 2 | 196 | |||
| GenBank mRNA | BC047546.1 | - | 1, 2 | 193 | ||
| BG527749.1 | - | 1, 2 | 69 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENTPD3-AS1| Gene Symbol: ENTPD3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040100.1 | 1, 2 | 196 | |||
| GenBank mRNA | BC047546.1 | - | 1, 2 | 193 | ||
| BG527749.1 | - | 1, 2 | 69 | |||