siRNA Details
| Gene Symbol: LOC100287728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW450913.1 | - | 1 | 43 | ||
| Gene Symbol: ZNF75D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI264634.1 | - | 1 | 378 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000416873.1 | - | 1 | 30 | Chr. X - 135123239 - 135123604 [-] on Build GRCh38 | |

LOC100287728
Genomic Map