siRNA Details
| Gene Symbol: LINCR-0002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120606.1 | 7 | 918 | |||
| GenBank mRNA | BX365943.2 | - | 4 | 636 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439804.5 | - | 4 | 637 | Chr. 3 - 191557324 - 191590356 [+] on Build GRCh38 | |

LINCR-0002
Genomic Map