siRNA Details
| Gene Symbol: LOC102723377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120672.1 | 3 | 343 | |||
| GenBank mRNA | BX103495.1 | - | 3 | 344 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428918.1 | - | 3 | 296 | Chr. 10 - 71508153 - 71511873 [-] on Build GRCh38 | |

LOC102723377
Genomic Map